Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TNF-alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
$241.00 - $549.50
Specifications
Antigen | TNF-alpha |
---|---|
Clone | 28401R |
Dilution | Neutralization 0.01-0.04 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Neutralization, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB610RSP
![]() |
R&D Systems
MAB610RSP |
25 μg |
Each for $241.00
|
|
|||||
MAB610R100
![]() |
R&D Systems
MAB610R100 |
100 μg |
Each for $549.50
|
|
|||||
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
TNF-alpha | |
Neutralization 0.01-0.04 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
P01375 | |
7124 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Detects human TNF-alpha in ELISAs and Western blots. In sandwich ELISAs, less than 0.05% cross-reactivity with recombinant human TNF-beta , recombinant mouse TNF-alpha , recombinant rat TNF-alpha , and recombinant porcine TNF-alpha is observed. |
28401R | |
Neutralization, ELISA | |
Unconjugated | |
Mouse | |
Human | |
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
TNF | |
Primary | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title