Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TNF-alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2103100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2103100 100 μg
MAB2103SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2103100 Supplier R&D Systems Supplier No. MAB2103100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen TNF-alpha
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 2342C
Conjugate Unconjugated
Dilution Western Blot 2 μg;/mL, Neutralization 1-10 ng/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P01375
Gene Alias APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Host Species Rabbit
Immunogen E. coli-derived recombinant human TNF-alpha , Val77-Leu233, Accession No. P01375
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7124.0
Test Specificity Detects human TNF-alpha in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.