Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human TNF-alpha Magnetic Luminex™ Performance Assay
Click to view available options
Quantity:
1 Kit
Description
- Magnetic format allows for easier wash steps
- Undergoes similar validation testing as Quantikine™ ELISA Kits
- Precise and Specific
- Requires a small sample volume (<50 μL)
- Run your assay in just 3-5 hours
- Mass-calibrated standards for consistent results with every new lot of material
- Enough reagents for one 96-well plate
Specifications
Specifications
Product Type | Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform |
Quantity | 1 Kit |
Species | Human |
Binding Target | TNF-alpha |
Bead Type | Magnetic |
Assay Sensitivity | 1.5pg/mL |
Assay Range | 6.00 - 4500pg/mL |
Gene ID (Entrez) | 7124 |
Gene Symbol | TNF |
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction