Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TNF-alpha (Research Grade Adalimumab Biosimilar) Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC9677N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC9677N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC9677N100 Supplier R&D Systems Supplier No. IC9677N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Monoclonal Antibody

TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen TNF-alpha
Applications Flow Cytometry
Classification Monoclonal
Clone Hu7
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry 0.25-1μg/106 cells
Formulation Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Host Species Human
Immunogen Human TNF-alpha
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7124
Test Specificity Detects human TNF-alpha in direct ELISAs. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Adalimumab. This product is for research use only.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.