Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TNF-alpha (Research Grade Adalimumab Biosimilar) Biotinylated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. IC9677B100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC9677B100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC9677B100 Supplier R&D Systems Supplier No. IC9677B100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Monoclonal Antibody

TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen TNF-alpha
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone Hu7
Conjugate Biotin
Dilution Intracellular Staining by Flow Cytometry 10μL/106 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Alias APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Host Species Human
Immunogen Human TNF-alpha
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 7124
Test Specificity Detects human TNF-alpha in direct ELISAs. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Adalimumab. This product is for research use only.
Target Species Human
Content And Storage Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.