Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TNF RI/TNFRSF1A Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 7 publications
Supplier: R&D Systems AF225SP
This item is not returnable.
View return policy
Description
TNF RI/TNFRSF1A Polyclonal specifically detects TNF RI/TNFRSF1A in Human samples. It is validated for Western Blot, Flow Cytometry, Agonist Activity, Immunocytochemistry, CyTOF-ready.Specifications
TNF RI/TNFRSF1A | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNFRI, TNF-RI, TNF-R-I, TNFR-I, TNFRSF1A, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Detects human sTNF RI/TNFRSF1A in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant mouse TNF sRI and recombinant rat TNF sRI is observed. This anti-human TNF RI antibody can interact with the cell surface | |
Human | |
IgG |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Agonist Activity 1-6 ug/mL, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
NP_001056 | |
TNFRSF1A | |
E. coli-derived recombinant human sTNF RI/TNFRSF1A Accession # NP_001056 | |
25 μg | |
Agonist Activity | |
7132 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction