Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TNF RI/TNFRSF1A Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 19 publications
Supplier: R&D Systems MAB625500
This item is not returnable.
View return policy
Description
TNF RI/TNFRSF1A Monoclonal specifically detects TNF RI/TNFRSF1A in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
TNF RI/TNFRSF1A | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
TNFRSF1A | |
E. coli-derived recombinant human TNF RI/TNFRSF1A Extracellular domain | |
500 μg | |
Primary | |
Detects human TNF RI/TNFRSF1A in ELISAs and Western blots. In sandwich immunoassays, no significant cross-reactivity or interference with recombinant human (rh) TGF-alpha , hTGF-beta 1, porcine (p) TGF-beta 1, rhTGF-beta 1, pTGF-beta 1.2, pTGF-beta 2, recombinant chicken TGF-beta 3, rhTGF-beta 3, recombinant amphibian TGF-beta 5, rhTNF-beta , or rhTNF RII is observed. | |
Human | |
Ascites |
Western Blot, Neutralization, ELISA | |
16805 | |
Western Blot 1 ug/mL, Neutralization 1-6 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNFRI, TNF-RI, TNF-R-I, TNFR-I, TNFRSF1A, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
7132 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction