Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TNF RI/TNFRSF1A PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 16 publications
Supplier: R&D Systems FAB225P
This item is not returnable.
View return policy
Description
TNF RI/TNFRSF1A Monoclonal specifically detects TNF RI/TNFRSF1A in Human samples. It is validated for Flow Cytometry.Specifications
TNF RI/TNFRSF1A | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNFRI, TNF-RI, TNF-R-I, TNFR-I, TNFRSF1A, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
7132 | |
Human | |
Purified |
Flow Cytometry | |
16803 | |
Flow Cytometry 10 uL/10^6 cells | |
NP_001056 | |
TNFRSF1A | |
E. coli-derived recombinant human TNF RI/TNFRSF1A Accession # NP_001056 | |
100 Tests | |
Primary | |
Detects human TNF RI in Western blots. In Western blots, does not cross-react with recombinant human TNF RII, recombinant mouse (rm) TNF RI, or rmTNF RII. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction