Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human TNF RII/TNFRSF1B ELISA Kit (Colorimetric)
SDP

Catalog No. NBP188105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP188105 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP188105 Supplier Novus Biologicals™ Supplier No. NBP188105

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human TNF RII/TNFRSF1B ELISA Kit (Colorimetric) measures human TNF RII/TNFRSF1B in cell culture supernatant, serum, plasma (edta, citrate, heparin), urine.

Specifications

Assay Range 0.16 - 10.0ng/mL
Assay Sensitivity 0.10ng/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 7133
Gene Symbol TNFRSF1B
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sTNF-R (80 kDa), HRP-Conjugate anti-human sTNF-R (80 kDa) monoclonal antibody, Human sTNF-R (80 kDa) Standard, 10ng/mL, Control, lyophilized, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Apoptosis, Cancer, Cytokine Research, Zinc Finger
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNF-RII, TNF-R-II, TNFR-II, tumor necrosis factor beta receptor, tumor necrosis factor bi
Target TNF RII/TNFRSF1B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.