Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TNFSF11 (O14788, 174 amino acids) Partial Recombinant Protein

Catalog No. 89965256
Click to view available options
Quantity:
10 μg

Used for Func, SDS-PAGE

This gene encodes a member of the tumor necrosis factor (TNF) cytokine family which is a ligand for osteoprotegerin and functions as a key factor for osteoclast differentiation and activation. This protein was shown to be a dentritic cell survival factor and is involved in the regulation of T cell-dependent immune response. T cell activation was reported to induce expression of this gene and lead to an increase of osteoclastogenesis and bone loss. This protein was shown to activate antiapoptotic kinase AKT/PKB through a signaling complex involving SRC kinase and tumor necrosis factor receptor-associated factor (TRAF) 6, which indicated this protein may have a role in the regulation of cell apoptosis. Targeted disruption of the related gene in mice led to severe osteopetrosis and a lack of osteoclasts. The deficient mice exhibited defects in early differentiation of T and B lymphocytes, and failed to form lobulo-alveolar mammary structures during pregnancy. Two alternatively spliced transcript variants have been found. [provided by RefSeq]

Specifications

Accession Number O14788
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 8600
Molecular Weight (g/mol) 20kDa
Name TNFSF11 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 10 μg
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with 5 mM Tris, pH 7.5, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg
Gene Alias CD254/ODF/OPGL/OPTB2/RANKL/TRANCE/hRANKL2/sOdf
Common Name TNFSF11
Gene Symbol TNFSF11
Biological Activity Determined by its ability to neutralize the stimulation of U937 cells treated with 10ng/mL of sRANKL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade ≥98% by SDS-PAGE and HPLC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.