Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TOB1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5228
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5228 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5228 Supplier R&D Systems Supplier No. AF5228
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

TOB1 Polyclonal specifically detects TOB1 in Human samples. It is validated for Western Blot.

Specifications

Antigen TOB1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Gene Accession No. P50616
Gene Alias APRO6, MGC104792, PIG49, protein Tob1, TOBMGC34446, Transducer of erbB-2 1, transducer of ERBB2, 1, TROB, TROB1
Gene Symbols TOB1
Host Species Goat
Immunogen E. coli-derived recombinant human TOB1 Met1-Asn345 Accession # P50616
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10140
Test Specificity Detects endogenous human TOB1 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.