Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Tollip Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4678
This item is not returnable.
View return policy
Description
Tollip Polyclonal specifically detects Tollip in Human samples. It is validated for Western Blot.Specifications
Tollip | |
Polyclonal | |
Unconjugated | |
Q9H0E2 | |
TOLLIP | |
E. coli-derived recombinant human Tollip Met1-Pro274 Accession # Q9H0E2 | |
100 μg | |
Primary | |
Detects endogenous human Tollip in Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
adapter protein, FLJ33531, IL-1RAcPIP, toll interacting protein, toll-interacting protein | |
Sheep | |
Affinity Purified | |
RUO | |
54472 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction