Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Total DDR1 DuoSet IC ELISA
SDP

Catalog No. DYC23965 Shop All Novus Biologicals Products
Change view
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DYC23965 5 Plates
DYC23962 2 Plates
DYC2396E 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC23965 Supplier R&D Systems™ Supplier No. DYC23965

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains the basic components required for the development of sandwich ELISAs to measure in cell lysates.

The Human Total DDR1 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human DDR1.

Specifications

Product Type ELISA Development Kit for Intracellular Proteins
Quantity 1 Kit
Species Human
Conjugate Biotin
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 780
Product Line R&D Systems
Packaging Quantity 5 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Signaling
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.