Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Total ERK1 DuoSet IC ELISA
SDP

Catalog No. DYC19405 Shop All Novus Biologicals Products
Change view
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DYC19405 5 Plates
DYC19402 2 Plates
DYC1940E 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC19405 Supplier R&D Systems™ Supplier No. DYC19405

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Total ERK1 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Total ERK1 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human ERK1.

Specifications

Product Type ELISA Development Kit for Intracellular Proteins
Quantity 1 Kit (for 5 Plates)
Species Human
Conjugate Biotin
Assay Range 312.00 to 20000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 5595
Product Line R&D Systems
Synonym EC 2.7.11, ERK-1, ERK1p44-MAPK, ERT2, Extracellular signal-regulated kinase 1, extracellular signal-related kinase 1, HS44KDAP, HUMKER1A, Insulin-stimulated MAP2 kinase, MAP kinase 1, MAP kinase 3, MAP kinase isoform p44, MAPK 1, MAPK 3, MAPK3, MGC20180, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 1, mitogen-activated protein kinase 3, p44erk1, p44-ERK1, P44MAPK, PRKM3, PRKM3EC 2.7.11.24
Packaging Quantity 5 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Signaling
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.