Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Total Mer DuoSet IC ELISA
SDP

Catalog No. DYC8912 Shop All R&D Systems Products
Change view
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DYC8912 2 Plates
DYC8915 5 Plates
DYC891E 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC8912 Supplier R&D Systems™ Supplier No. DYC8912
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Total Mer DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Complete ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. Kits are available for detection and enumeration of a single analyte or two analytes simultaneously.

The Human Total Mer DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human Mer.

Specifications

Product Type ELISA Development Kit for Intracellular Proteins
Quantity 1 Kit
Species Human
Conjugate Biotin
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 10461
Packaging Quantity 2 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.