Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Total MMP-3 Quantikine ELISA Kit

Catalog No. p-4753721
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 Plate
This item is not returnable. View return policy
This item is not returnable. View return policy

Human Total MMP-3 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Total MMP-3 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human MMP-3 in cell culture supernates (50 ul), serum (10 ul), heparin plasma (10 ul).

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.045 ng/mL
Conjugate HRP
Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (Heparin), Serum
Specificity Natural and Recombinant Human total MMP-3
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 4314
Gene Symbol MMP3
Quantity 1 Plate
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP3, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, Stromelysin 1, stromelysin-1, Transin-1
Target MMP-3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.