Learn More
R&D Systems™ Human Total MSP R/Ron DuoSet IC
Description
- Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
- Development protocols are provided to guide further assay optimization
- Assay can be customized to your specific needs
- Economical alternative to complete kits
Specifications
Specifications
| Product Type | ELISA Development Kit for Intracellular Proteins |
| Quantity | 1 Kit |
| Species | Human |
| Conjugate | Biotin |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 4486 |
| Packaging Quantity | 5 Plates |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.