Learn More
R&D Systems™ Human Total p27/Kip1 DuoSet IC ELISA
Description
Specifications
Specifications
| Product Type | ELISA Development Kit for Intracellular Proteins |
| Quantity | 1 Kit |
| Species | Human |
| Conjugate | Biotin |
| Assay Range | 62.50 to 4000 pg/mL |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 1027 |
| Packaging Quantity | 2 Plates |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.