Learn More
Gibco™ Human TPO (Thrombopoietin), Animal-Free Recombinant Protein, PeproTech®
Description
Recombinant Human TPO is a fully biologically active 174 amino acid polypeptide (18.6 kDa), which contains the erythropoietin-like domain of the full length TPO protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P40225 |
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Format | Lyophilized |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 7066 |
| Molecular Weight (g/mol) | 18.6 kDa |
| Name | Human TPO (Thrombopoietin), Animal-Free |
| Purification Method | HPLC, SDS-PAGE |
| Quantity | 250 μg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.