Learn More
Human TR beta 1/NR1A2 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | TR beta 1/NR1A2/Thyroid Hormone Receptor beta |
| Applications | Western Blot, Immunoprecipitation, ELISA |
| Classification | Monoclonal |
| Clone | H3825A |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunoprecipitation, Direct ELISA |
| Formulation | A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide |
| Gene Alias | avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 2, c-erbA-2, c-erbA-beta, ERBA2MGC126109, ERBA-BETA, GRTH, MGC126110, NR1A2, NR1A2thyroid hormone receptor, beta (avian erythroblastic leukemia viral (v-erb-a)oncogene homolog 2), Nuclear receptor subfamily 1 group A member 2, oncogene ERBA2, PRTH, THR1pituitary resistance to thyroid hormone, THRB, THRB1, THRB2, thyroid hormone receptor beta, thyroid hormone receptor beta 1, thyroid hormone receptor, beta (erythroblastic leukemia viral (v-erb-a)oncogene homolog 2, avian), v-ErbA2 |
| Gene Symbols | THRB |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.