Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TR4/NR2C2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. PPH0107B00
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPH0107B00 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPH0107B00 Supplier R&D Systems Supplier No. PPH0107B00
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TR4/NR2C2 Monoclonal specifically detects TR4/NR2C2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Direct ELISA.

Specifications

Antigen TR4/NR2C2
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, ELISA
Classification Monoclonal
Clone H0107B
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Direct ELISA
Formulation A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide
Gene Alias hTAK1, NR2C2, Nuclear hormone receptor TR4, nuclear receptor subfamily 2 group C member 2, nuclear receptor subfamily 2, group C, member 2, Orphan nuclear receptor TAK1, Orphan nuclear receptor TR4, TAK1, TAK1TR4TR2R1, Testicular receptor 4, TR4 nuclear hormone receptor
Gene Symbols NR2C2
Host Species Mouse
Immunogen Recombinant human TR4/NR2C2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7182
Test Specificity This antibody specifically recognizes human TR4 and cross-reacts with mouse and rat TR4. Not yet tested in other species.
Target Species Human
Content And Storage This antibody is stable for greater than six months when held at -20 degreesC in a manual defrost freezer or at -70 degreesC. Upon thawing, the antibody can be stored at 2-8 degreesC for at least 1 month without detectable loss of activity. Avoid repeated freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.