Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TRA-1-60(R) Neuraminidase Resistant Epitope Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB4770T100UG
This item is not returnable.
View return policy
Description
TRA-1-60(R) Monoclonal specifically detects TRA-1-60(R) in Human samples. It is validated for Flow Cytometry.Specifications
TRA-1-60(R) | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
GCTM, PCLP, POD XL, PODXL, RA160, TRA160(R), tra61 | |
Mouse | |
100 μg | |
Primary | |
Human | |
IgM |
Flow Cytometry | |
TRA-1-60 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
PODXL | |
2102Ep human embryonal carcinoma cell line | |
RUO | |
Detects human TRA-1-60(R). | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction