Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TRA-1-81 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8495U100UG
This item is not returnable.
View return policy
Description
TRA-1-81 Monoclonal specifically detects TRA-1-81 in Human samples. It is validated for Flow Cytometry.Specifications
TRA-1-81 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Mouse | |
100 μg | |
Primary | |
Human | |
IgM |
Flow Cytometry | |
TRA-1-81 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
GCTM, gp200, MGC138240, PC, PCLP, POD XL, PODXL, TRA181 | |
Human embryonal carcinoma cell line 2102Ep | |
RUO | |
Detects human TRA-1-81 in flow cytometry. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction