Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TRA-1-85/CD147 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3195N
This item is not returnable.
View return policy
Description
TRA-1-85/CD147 Monoclonal specifically detects TRA-1-85/CD147 in Human samples. It is validated for Flow Cytometry.Specifications
TRA-1-85/CD147 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Detects human TRA-1-85 antigen in flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry | |
TRA-1-85 | |
Flow Cytometry 5 uL/10^6 cells | |
BSG | |
2120Ep human embryonal carcinoma cell line | |
100 Tests | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction