Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TRADD Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC2658N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC2658N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC2658N100 Supplier R&D Systems Supplier No. IC2658N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRADD Monoclonal specifically detects TRADD in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen TRADD
Applications Flow Cytometry
Classification Monoclonal
Clone 313203
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q15628
Gene Alias MGC11078, TNFRSF1A-associated via death domainTNFR1-associated DEATH domain protein, tumor necrosis factor receptor type 1 associated death domain protein, tumor necrosis factor receptor type 1-associated DEATH domain protein, tumor necrosis factor receptor-1-associated protein
Gene Symbols TRADD
Host Species Mouse
Immunogen E. coli-derived recombinant human TRADD Met1-Ala312 Accession # Q15628
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8717
Test Specificity Detects human TRADD in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) BAFF R, rhTNF R1, rhHVEM, rhLT beta R, rhCD30, rhGITR, rh4-1BB, rhDR3, rhDR6, rhRANK, rhRELT, rhTAJ, rhEDAR, rhFas, recombinant mouse (rm) DC-TRAIL R1, rmDC-TRAIL R2, rhTRAIL R3, rhTRAIL R4, rhNGF R, rhCD27, rhTNF RII, rhTWEAK R, rhTRAIL R2, or rhCD40 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.