Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TRAF-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3279
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3279 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3279 Supplier R&D Systems Supplier No. MAB3279
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

TRAF-4 Monoclonal specifically detects TRAF-4 in Human samples. It is validated for Western Blot.

Specifications

Antigen TRAF-4
Applications Western Blot
Classification Monoclonal
Clone 433709
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9BUZ4
Gene Alias CART1, CART1RING finger protein 83, Cysteine-rich domain associated with RING and Traf domains protein 1, Metastatic lymph node gene 62 protein, MLN 62, MLN62, MLN62cysteine-rich domain associated with ring and TRAF domain, RNF83malignant 62, TNF receptor-associated factor 4, TRAF4, tumor necrosis receptor-associated factor 4A
Gene Symbols TRAF4
Host Species Rat
Immunogen E. coli-derived recombinant human TRAF-4 Met1-Ser470 Accession # Q9BUZ4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9618
Test Specificity Detects endogenous human TRAF-4. In Western blots, this antibody does not cross-react with rhTRAF-1, -2, -3, -5, or -6 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.