Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TRAF-5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3280SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3280SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3280SP Supplier R&D Systems Supplier No. AF3280SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

TRAF-5 Polyclonal specifically detects TRAF-5 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen TRAF-5
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O00463
Gene Alias MGC:39780, RNF84, RNF84RING finger protein 84, TNF receptor-associated factor 5, TRAF5
Gene Symbols TRAF5
Host Species Sheep
Immunogen E. coli-derived recombinant human TRAF-5 Lys353-Leu557 Accession # O00463
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7188
Test Specificity Detects human TRAF-5 in direct ELISAs. In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) TRAF-3 is observed, and less than 5% cross-reactivity with rhTRAF-2 and rhTRAF-4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.