Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TRAF-6 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB3284N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3284N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3284N100 Supplier R&D Systems Supplier No. FAB3284N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRAF-6 Monoclonal specifically detects TRAF-6 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen TRAF-6
Applications Flow Cytometry
Classification Monoclonal
Clone 326019
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q9Y4K3
Gene Alias E3 ubiquitin-protein ligase TRAF6, EC 6.3.2.-, Interleukin-1 signal transducer, RING finger protein 85, RNF85MGC:3310, TNF receptor-associated factor 6, TRAF6
Host Species Mouse
Immunogen E.coli-derived recombinant human TRAF-6 Met1-Val522 Accession # Q9Y4K3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7189
Test Specificity Detects human TRAF-6 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.