Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TRAIL R2/TNFRSF10B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6313
This item is not returnable.
View return policy
Description
TRAILR2/TNFRSF10B Monoclonal specifically detects TRAILR2/TNFRSF10B in Human samples. It is validated for Western Blot.Specifications
TRAILR2/TNFRSF10B | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
NP_003833 | |
TNFRSF10B | |
E. coli-derived recombinant human TRAIL R2/TNFRSF10B Ile56-Pro128 Accession # NP_003833 | |
100 μg | |
Primary | |
Detects human TRAIL R2/TNFRSF10B in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) 4-1BB, rhBAFF R, rhCD27, rhCD30, rhCD40, rhDR3, rhDR6, rhEDAR, rhFas, rhGITR, rhHVEM, rhOPG, rhRANK, rhRELT, rhTNF RI, rhTNF RII, rhTRAIL R1, rhTRAIL R3, rhTRAIL R4, or rhTWEAK R is observed. Additionally in direct ELISAs, no cross-reactivity with rhTRAIL R4, rhXEDAR, recombinant mouse (rm) DR5, rhTRAIL R1, rmTRAIL R2, or rmTNFRH3 is observed. | |
Human | |
Purified |
Western Blot | |
603307 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TNFRSF10B, TRAIL receptor 2, TRAILR2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8795 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction