Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TRAIL R2/TNFRSF10B Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 8 publications

$200.50 - $2020.00

Specifications

Antigen TRAILR2/TNFRSF10B
Clone 71903
Dilution Agonist Activity 2-12 ng/mL
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB631SP
GREENER_CHOICE
View Documents
R&D Systems
MAB631SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB631100
GREENER_CHOICE
View Documents
R&D Systems
MAB631100
100 μg
Each for $549.50
Only null left
Add to Cart
 
MAB631500
GREENER_CHOICE
View Documents
R&D Systems
MAB631500
500 μg
Each for $2,020.00
Only null left
Add to Cart
 
Description

Description

TRAILR2/TNFRSF10B Monoclonal specifically detects TRAILR2/TNFRSF10B in Human samples. It is validated for Agonist Activity.
Specifications

Specifications

TRAILR2/TNFRSF10B
Agonist Activity 2-12 ng/mL
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TNFRSF10B, TRAIL receptor 2, TRAILR2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9
TNFRSF10B
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
71903
Monoclonal
Mouse
Human
CAG46696
8795
Mouse myeloma cell line NS0-derived recombinant human TRAIL R2/TNFRSF10B Met1-Glu182 Accession # CAG46696
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human TRAIL R2/TNFRSF10B in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) TRAIL R1, rhTRAIL R3, rhTRAIL R4, or rhDcR3.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.