Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TRAIL R3/TNFRSF10C Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6302S100UG
This item is not returnable.
View return policy
Description
TRAILR3/TNFRSF10C Monoclonal specifically detects TRAILR3/TNFRSF10C in Human samples. It is validated for Flow Cytometry.Specifications
TRAILR3/TNFRSF10C | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
O14798 | |
TNFRSF10C | |
Mouse myeloma cell line NS0-derived recombinant human TRAIL R3/TNFRSF10C Met1-Ala221 Accession # O14798 | |
RUO | |
8794 | |
Human | |
IgG1 |
Flow Cytometry | |
90906 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Antagonist decoy receptor for TRAIL/Apo-2L, CD263, CD263 antigen, cytotoxic TRAIL receptor-3, DcR1, DCR1MGC149502, Decoy receptor 1, Decoy TRAIL receptor without death domain, LITMGC149501, Lymphocyte inhibitor of TRAIL, TNFRSF10C, TRAIL receptor 3, TRAIL receptor without an intracellular domain, TRAILR3, TRAIL-R3, TRAILR3TNF-related apoptosis-inducing ligand receptor 3, TRIDDCR1-TNFR, tumor necrosis factor receptor superfamily member 10C, tumor necrosis factor receptor superfamily, member 10c, decoy without anintracellular domain | |
Mouse | |
100 μg | |
Primary | |
Detects human TRAIL R3/TNFRSF10C in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) TRAIL R2, rhTRAIL R1, or rhTRAIL R4 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction