Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TRAILR4/TNFRSF10D Alexa Fluor 350 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB633U100UG
This item is not returnable.
View return policy
Description
TRAILR4/TNFRSF10D/DcR2 Monoclonal specifically detects TRAILR4/TNFRSF10D/DcR2 in Human samples. It is validated for Flow Cytometry.Specifications
TRAILR4/TNFRSF10D/DcR2 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9UBN6 | |
TNFRSF10D | |
Mouse myeloma cell line NS0-derived recombinant human TRAIL R4/TNFRSF10D Ala56-His211 Accession # Q9UBN6 | |
RUO | |
8793 | |
Human | |
IgG1 |
Flow Cytometry | |
104918 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD264, CD264 antigen, DcR2, DCR2decoy with truncated death domain, Decoy receptor 2, TNF receptor-related receptor for TRAIL, TNF-related apoptosis-inducing ligand receptor 4, TNFRSF10D, TRAIL R4, TRAIL-R4, TRAILR4TRAIL receptor 4, TRUNDDTRAIL receptor with a truncated death domain, tumor necrosis factor receptor superfamily member 10D, tumor necrosis factor receptor superfamily, member 10d, decoy with truncateddeath domain | |
Mouse | |
100 μg | |
Primary | |
Detects human TRAIL R4/TNFRSF10D in ELISAs and Western blots. In sandwich immunoassays, less than 5% cross-reactivity with recombinant human (rh) TRAIL R1, rhTRAIL R2, TRAIL R3, rhTRAIL, rhTNF-alpha , and rhTNF-beta is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction