Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TRAILR4/TNFRSF10D Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB633R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB633R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB633R100 Supplier R&D Systems Supplier No. FAB633R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRAILR4/TNFRSF10D/DcR2 Monoclonal specifically detects TRAILR4/TNFRSF10D/DcR2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TRAILR4/TNFRSF10D/DcR2
Applications Flow Cytometry
Classification Monoclonal
Clone 104918
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9UBN6
Gene Alias CD264, CD264 antigen, DcR2, DCR2decoy with truncated death domain, Decoy receptor 2, TNF receptor-related receptor for TRAIL, TNF-related apoptosis-inducing ligand receptor 4, TNFRSF10D, TRAIL R4, TRAIL-R4, TRAILR4TRAIL receptor 4, TRUNDDTRAIL receptor with a truncated death domain, tumor necrosis factor receptor superfamily member 10D, tumor necrosis factor receptor superfamily, member 10d, decoy with truncateddeath domain
Gene Symbols TNFRSF10D
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TRAIL R4/TNFRSF10D Ala56-His211 Accession # Q9UBN6
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8793
Test Specificity Detects human TRAIL R4/TNFRSF10D in ELISAs and Western blots. In sandwich immunoassays, less than 5% cross-reactivity with recombinant human (rh) TRAIL R1, rhTRAIL R2, TRAIL R3, rhTRAIL, rhTNF-alpha , and rhTNF-beta is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.