Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TRANCE/TNFSF11/RANK L Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6264R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6264R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6264R100 Supplier R&D Systems Supplier No. FAB6264R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRANCE/TNFSF11/RANK L Monoclonal specifically detects TRANCE/TNFSF11/RANK L in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TRANCE/TNFSF11/RANK L
Applications Flow Cytometry
Classification Monoclonal
Clone 685857
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide
Gene Accession No. O14788
Gene Alias CD254, CD254 antigen, ODF, OPGL, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANK L, RANKL, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TNFSF11, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TRANCE/TNFSF11/RANK L, Gly136-Asp317, Accession No. O14788
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 8600
Test Specificity Detects human TRANCE/TNFSF11/RANK L in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.