Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TRANCE/TNFSF11/RANK L Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
$214.00 - $2020.00
Specifications
| Antigen | TRANCE/TNFSF11/RANK L |
|---|---|
| Clone | 70525 |
| Dilution | Immunohistochemistry 8-25 ug/mL, Neutralization 0.8-2.5 ug/mL |
| Applications | Immunohistochemistry, Neutralization |
| Classification | Monoclonal |
Description
TRANCE/TNFSF11/RANK L Monoclonal specifically detects TRANCE/TNFSF11/RANK L in Human samples. It is validated for Immunohistochemistry, Neutralization.Specifications
| TRANCE/TNFSF11/RANK L | |
| Immunohistochemistry 8-25 ug/mL, Neutralization 0.8-2.5 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD254, CD254 antigen, ODF, OPGL, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANK L, RANKL, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TNFSF11, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
| TNFSF11 | |
| IgG2b | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 70525 | |
| Immunohistochemistry, Neutralization | |
| Unconjugated | |
| Mouse | |
| Human | |
| O14788 | |
| 8600 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human TRANCE/TNFSF11/RANK L Gly136-Asp317 (Ala194Gly) Accession # O14788 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human TRANCE/TNFSF11/RANK L in direct ELISAs. No cross-reactivity with recombinant mouse TRANCE is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title