Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Transcobalamin II Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7895
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7895 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7895 Supplier R&D Systems Supplier No. MAB7895
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Transcobalamin II Monoclonal specifically detects Transcobalamin II in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Transcobalamin II
Applications Western Blot
Classification Monoclonal
Clone 874420
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Simple Western 20 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P20062
Gene Alias D22S676, D22S750, macrocytic anemia, TC, TC-2, TC2II, TCII, TCN2, transcobalamin II; macrocytic anemia, transcobalamin IITC II, transcobalamin-2, vitamin B12-binding protein 2
Gene Symbols TCN2
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Transcobalamin II Glu19-Trp427 Accession # P20062
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6948
Test Specificity Detects human Transcobalamin II in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.