Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Transferrin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2914100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2914100 100 μg
AF2914SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2914100 Supplier R&D Systems Supplier No. AF2914100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Antibody

Transferrin specifically detects Transferrin in Human samples. It is validated for Western Blot.

Specifications

Antigen Transferrin
Applications Western Blot
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Alias ApoTransferrin, Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, HoloTransferrin, PRO1557, PRO2086, serotransferrin, Siderophilin, TF, transferrin
Host Species Goat
Immunogen Human plasma-derived
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7018
Test Specificity Detects Human Transferrin in ELISAs. Detects human Transferrin and Apo-Transferrin in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.