Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Transglutaminase 3/TGM3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4604SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4604SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4604SP Supplier R&D Systems Supplier No. AF4604SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Transglutaminase 3/TGM3 Polyclonal specifically detects Transglutaminase 3/TGM3 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Transglutaminase 3/TGM3
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_003236
Gene Alias EC 2.3.2.13, MGC126250, protein-glutamine gamma-glutamyltransferase E, TG(E), TG3, TGase E, TGase-3, TgaseE, TGEMGC126249, TGM3, transglutaminase 3 (E polypeptide, protein-glutamine-gamma-glutamyltransferase), Transglutaminase E, transglutaminase-3
Gene Symbols TGM3
Host Species Sheep
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Transglutaminase 3/TGM3, Ala2-Glu693, Accession # NP_003236
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7053
Test Specificity Detects human Transglutaminase 3/TGM3 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.