Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Transthyretin/Prealbumin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7505SP
This item is not returnable.
View return policy
Description
Transthyretin/Prealbumin Monoclonal specifically detects Transthyretin/Prealbumin in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Transthyretin/Prealbumin | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
ATTR, HsT2651, PALB, PALBTBPA, Prealbumin, prealbumin, amyloidosis type I, TBPA, thyroxine-binding prealbumin, transthyretin, TTR | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7276 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Immunocytochemistry | |
760430 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
TTR | |
Human Transthyretin/Prealbumin | |
25 μg | |
Primary | |
Detects human Transthyretin/Prealbumin in direct ELISAs and Western blots. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction