Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Trappin-2/Elafin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1747SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1747SP 25 μg
MAB1747 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1747SP Supplier R&D Systems Supplier No. MAB1747SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Trappin-2/Elafin/Skalp Monoclonal specifically detects Trappin-2/Elafin/Skalp in Human samples. It is validated for Western Blot.

Specifications

Antigen Trappin-2/Elafin/Skalp
Applications Western Blot
Classification Monoclonal
Clone 257729
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P19957
Gene Alias cementoin, Elafin, Elastase-specific inhibitor, ESI, ESIWAP four-disulfide core domain protein 14, Peptidase inhibitor 3, peptidase inhibitor 3, skin-derived, PI3, PI-3, pre-elafin, protease inhibitor 3, skin-derived (SKALP), Protease inhibitor WAP3, SKALP, SKALPELAFIN, Skin-derived antileukoproteinase, trappin-2, Trappin2, WAP four-disulfide core domain 14, WAP3MGC13613, WFDC14elafin
Gene Symbols PI3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Trappin-2/Elafin Ala23-Gln117 Accession # P19957
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5266
Test Specificity Detects the full length form (amino acids 23-117) and the trypsin-processed form (amino acids 61-117) of human Trappin-2/Elafin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human SLPI is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.