Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TREM-1 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1278R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1278R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1278R100 Supplier R&D Systems Supplier No. FAB1278R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TREM1 Monoclonal specifically detects TREM1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TREM1
Applications Flow Cytometry
Classification Monoclonal
Clone 193015
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9NP99
Gene Alias CD354, CD354 antigen, TREM1, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1
Gene Symbols TREM1
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human TREM-1 Ala21-Asn205 Accession # Q9NP99
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54210
Test Specificity Detects human TREM-1 in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.