Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TREM-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 13 publications
$214.00 - $2070.00
Specifications
Antigen | TREM1 |
---|---|
Clone | 193015 |
Concentration | LYOPH |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Agonist Activity, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1278SP
![]() |
R&D Systems
MAB1278SP |
25 μg |
Each for $214.00
|
|
|||||
MAB1278100
![]() |
R&D Systems
MAB1278100 |
100 μg |
Each for $573.00
|
|
|||||
MAB1278500
![]() |
R&D Systems
MAB1278500 |
500 μg |
Each for $2,070.00
|
|
|||||
Description
TREM1 Monoclonal specifically detects TREM1 in Human samples. It is validated for Flow Cytometry, Agonist Activity, CyTOF-ready.Specifications
TREM1 | |
LYOPH | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Agonist Activity | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD354, CD354 antigen, TREM1, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
TREM1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
193015 | |
Flow Cytometry 0.25 ug/10^6 cells, Agonist Activity, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q9NP99 | |
54210 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human TREM-1 Ala21-Asn205 Accession # Q9NP99 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human TREM-1 in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title