Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TREM-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $456.00
Specifications
Antigen | TREM1 |
---|---|
Clone | 888137 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB12782SP
![]() |
R&D Systems
MAB12782SP |
25 μg |
Each for $200.50
|
|
|||||
MAB12782100
![]() |
R&D Systems
MAB12782100 |
100 μg |
Each for $456.00
|
|
|||||
Description
TREM1 Monoclonal specifically detects TREM1 in Human samples. It is validated for ELISA.Specifications
TREM1 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
CD354, CD354 antigen, TREM1, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
E. coli-derived recombinant human TREM-1, Ala21-Asn150, Accession # Q9NP99 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
888137 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
Q9NP99 | |
54210 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title