Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TREM-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1278
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1278 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1278 Supplier R&D Systems Supplier No. AF1278
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

TREM1 Polyclonal specifically detects TREM1 in Human samples. It is validated for Western Blot, Flow Cytometry, Agonist Activity, CyTOF-ready, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen TREM1
Applications Western Blot, Flow Cytometry, Agonist Activity, CyTOF, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Agonist Activity, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Gene Accession No. Q9NP99
Gene Alias CD354, CD354 antigen, TREM1, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1
Gene Symbols TREM1
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human TREM-1 Met1-Arg200 Accession # Q9NP99
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54210
Test Specificity Detects human TREM-1 in ELISAs and Western blots. In ELISAs, less than 0.2% cross-reactivity with recombinant mouse (rm) TREM-1 and rmTREM-2b is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.