Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TREM-1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems BAF1278
This item is not returnable.
View return policy
Description
TREM1 Polyclonal specifically detects TREM1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA Detection (Matched Antibody Pair).Specifications
TREM1 | |
Polyclonal | |
Biotin | |
Q9NP99 | |
TREM1 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human TREM-1 Met1-Arg200 Accession # Q9NP99 | |
50 μg | |
Primary | |
Detects human TREM-1 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant mouse (rm) TREM-1 and rmTREM-2b is observed. | |
Human | |
IgG |
Western Blot, Flow Cytometry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
CD354, CD354 antigen, TREM1, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
Goat | |
Affinity Purified | |
RUO | |
54210 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction