Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TREM2 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB17291RU100UG
This item is not returnable.
View return policy
Description
TREM2 Monoclonal antibody specifically detects TREM2 in Human samples. It is validated for Flow CytometrySpecifications
TREM2 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
Mouse myeloma cell line NS0-derived recombinant mouse TREM2, Met1-Pro168, Accession # Q99NH8 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
237920R | |
Flow Cytometry | |
Q99NH8 | |
Rat | |
Protein A or G purified from cell culture supernatant | |
RUO | |
54209 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction