Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TREM2 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB17291RV1
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB17291RV1 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB17291RV1 Supplier R&D Systems Supplier No. FAB17291RV100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

TREM2 Monoclonal antibody specifically detects TREM2 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen TREM2
Applications Flow Cytometry
Classification Monoclonal
Clone 237920R
Conjugate Alexa Fluor 405
Dilution Flow Cytometry
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q99NH8
Gene Alias PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TREM2, Met1-Pro168, Accession # Q99NH8
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54209
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.