Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TREML1/TLT-1 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB2394V100UG
This item is not returnable.
View return policy
Description
TREML1/TLT-1 Monoclonal specifically detects TREML1/TLT-1 in Human samples. It is validated for Flow Cytometry.Specifications
TREML1/TLT-1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q86YW5 | |
TREML1 | |
Mouse myeloma cell line NS0-derived recombinant human TREML1/TLT-1 Gln16-Ser160 Accession # Q86YW5 | |
RUO | |
340205 | |
Human | |
IgG2b |
Flow Cytometry | |
268420 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
dJ238O23.3, GLTL1825, PRO3438, TLT1, TLT-1, TLT1MGC119173, trem-like transcript 1 protein, triggering receptor expressed on myeloid cells-like 1, Triggering receptor expressed on myeloid cells-like protein 1 | |
Rat | |
100 μg | |
Primary | |
Detects human TREML1/TLT-1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse TREML1 or recombinant human TREML2 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction