Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TRF-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5300SP
This item is not returnable.
View return policy
Description
TRF-1 Polyclonal specifically detects TRF-1 in Human samples. It is validated for Western Blot.Specifications
TRF-1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
NIMA-interacting protein 2, PIN-2, PIN2hTRF1-AS, Telomeric protein Pin2/TRF1, telomeric repeat binding factor (NIMA-interacting) 1, telomeric repeat binding factor 1, telomeric repeat binding protein 1, telomeric repeat-binding factor 1, TERF1, TRBF1, TRF1, TRF1FLJ41416, TRFt-TRF1, TTAGGG repeat-binding factor 1 | |
Sheep | |
Affinity Purified | |
RUO | |
7013 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
P54274 | |
TERF1 | |
E. coli-derived recombinant human TRF-1 Lys136-Lys295 Accession # P54274 | |
25 μg | |
Primary | |
Detects human TRF-1 in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction