Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TRF2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5635
This item is not returnable.
View return policy
Description
TRF-2 Polyclonal specifically detects TRF-2 in Human samples. It is validated for Western Blot.Specifications
TRF-2 | |
Polyclonal | |
Unconjugated | |
Q15554 | |
TERF2 | |
E. coli-derived recombinant human TRF2 Gln78-Thr238 Accession # Q15554 | |
100 μg | |
Primary | |
Detects endogenous human TRF2 in Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 2 ug/mL | |
Telomeric DNA-binding protein, telomeric repeat binding factor 2, Telomeric repeat binding protein 2, telomeric repeat-binding factor 2, TERF 2, TERF2, TRBF 2, TRBF2, TRF 2, TRF2, TTAGGG repeat binding factor 2, TTAGGG repeat-binding factor 2 | |
Goat | |
Affinity Purified | |
RUO | |
7014 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction